missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL18 (MIP-4) Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Description
Recombinant Human MIP-4 is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Macrophage Inflammatory Protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta and bone marrow. Although the MIP-4s receptor remains undetermined, MIP-4 is known to act as a chemoattractant for activated and non-activated T cells.
Specifications
Specifications
| Accession Number | P55774 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 6362 |
| Molecular Weight (g/mol) | 7.8 kDa |
| Name | Human CCL18 (MIP-4) |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction