missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL17 (TARC) Recombinant Protein, PeproTech®

Product Code. 17852493
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 17852493

Brand: Gibco™ 300305UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TARC is an 8.0 kDa protein containing 71 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92583
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6361
Molecular Weight (g/mol) 8 kDa
Name Human CCL17 (TARC)
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine
Common Name TARC
Gene Symbol CCL17
Biological Activity Determined by its ability to chemoattract human T cells using a concentration range of 1.0-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.