missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL17/TARC DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
Pack of 5
Description
The Human CCL17/TARC DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CCL17/TARC.
Specifications
Specifications
| Assay Range | 7.81 to 500 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| Gene ID (Entrez) | 6361 |
| Gene Symbol | CCL17 |
| Host Species | Human |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction