missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL17/TARC Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
£170.00 - £1855.00
Specifications
| Antigen | CCL17/TARC |
|---|---|
| Clone | 54026 |
| Dilution | Neutralization 0.4-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777148
![]() |
R&D Systems
MAB364-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18778729
![]() |
R&D Systems
MAB364-100 |
100 μg |
£464.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18121381
![]() |
R&D Systems
MAB364-500 |
500 μg |
£1855.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL17/TARC Monoclonal specifically detects CCL17/TARC in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CCL17/TARC | |
| Neutralization 0.4-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q92583.1 | |
| 6361 | |
| E. coli-derived recombinant human CCL17/TARC Ala24-Ser94 Accession # Q92583.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CCL17/TARC in ELISAs. Does not cross-react with recombinant human (rh) CCL2, 3, 4, 5, 7, 8, 11, 13, 16, 18, 19, 20, 21, 22, 23, 24, 25, rmCCL2, 3, 4, 5, 7, 9/10/MIP-1 gamma , 11, 12, 21, 22, 25, or rrCCL20. |
| 54026 | |
| Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
| CCL17 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title