missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCL16 (O15467, 24 a.a. - 120 a.a.) Partial Recombinant Protein

Product Code. 16201930
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 16201930

Brand: Abnova™ P3620.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10. [provided by RefSeq]

Sequence: QPKVPEWVNTPSTCCLKYYEKVLPRRLVVGYRKALNCHLPAIIFVTKRNREVCTNPNDDWVQEYIKDPNLPLLPTRNLSTVKIITAKNGQPQLLNSQ

Specifications

Accession Number O15467
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 6360
Molecular Weight (g/mol) 11kDa
Name CCL16 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 25 μg
Immunogen QPKVPEWVNTPSTCCLKYYEKVLPRRLVVGYRKALNCHLPAIIFVTKRNREVCTNPNDDWVQEYIKDPNLPLLPTRNLSTVKIITAKNGQPQLLNSQ
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Gene Alias CKb12/HCC-4/ILINCK/LCC-1/LEC/LMC/MGC117051/Mtn-1/NCC-4/NCC4/SCYA16/SCYL4
Common Name CCL16
Gene Symbol CCL16
Biological Activity Determined by its ability to chemoattract human monocytes using a concentration range of 2.0-40.0ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.