missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL14/HCC-1/HCC-3 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB324
This item is not returnable.
View return policy
Description
CCL14/HCC-1/HCC-3 Monoclonal specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for Western Blot.
Specifications
| CCL14/HCC-1/HCC-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 14, CCL14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6358 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 256413 | |
| Western Blot 1 ug/mL | |
| NP_116738 | |
| CCL14 | |
| E. coli-derived recombinant human CCL14/HCC-1/HCC-3 Thr20-Asn109 Accession # NP_116738 | |
| 500 μg | |
| Primary | |
| Detects human CCL14/HCC-1/HCC-3 in direct ELISAs and Western blots. Detects both CCL14a and CCL14b. In Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 28, recombinant mouse CCL2, 3, 4, 6, 7, 9/10/MIP-1 gamma , 11, 12, 19, 20, 21, 22, 24, 25, 28, or recombinant rat CCL20 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction