missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCK-AR Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2680N-100UG
This item is not returnable.
View return policy
Description
CCK-AR Monoclonal specifically detects CCK-AR in Human samples. It is validated for Flow Cytometry.
Specifications
| CCK-AR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P32238 | |
| CCKAR | |
| NS0 mouse myeloma cell line transfected with human CCK-A R Met1-Gln428 Accession # P32238 | |
| RUO | |
| 886 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 377251 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CCK1R, CCK1-R, CCK-A, CCK-A R, CCK-A receptor, CCKAR, CCK-AR, CCKRA, cholecystokinin A receptor, cholecystokinin receptor type A, cholecystokinin type-A receptor, Cholecystokinin-1 receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CCK-A R. Stains human CCK-A R transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only (RUO)
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido