missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CCDC48 (aa 438-511) Control Fragment Recombinant Protein

Product Code. 30209862
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209862

Brand: Invitrogen™ RP100120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (64%), Rat (64%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62261 (PA5-62261. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HA90
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79825
Name Human CCDC48 (aa 438-511) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C3orf73; Ccdc48; Ccdc48l1; coiled-coil domain containing 48; coiled-coil domain containing 48 like; coiled-coil domain containing 48-like1; coiled-coil domain-containing protein 48; EFCC1; EF-hand and coiled-coil domain containing 1; EF-hand and coiled-coil domain-containing protein 1; EF-hand domain-containing protein ENSP00000381169; EF-hand domain-containing protein ENSP00000381169 homolog
Common Name CCDC48
Gene Symbol EFCC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MTYFGHFGGANHAHTLGELEACIAMLVEQLRTQGCGGRTLGTSEEEAELQQKVEENEHLRLELQMVETERVRLS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.