missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCDC146 (aa 698-814) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54149 (PA5-54149. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function ofCCDC146 remains unknown.
Specifications
Specifications
| Accession Number | Q8IYE0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57639 |
| Name | Human CCDC146 (aa 698-814) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4930528G09Rik; Ccdc146; coiled-coil domain containing 146; coiled-coil domain-containing protein 146; KIAA1505; mKIAA1505 |
| Common Name | CCDC146 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction