missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CAZIP Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
CAZIP Synthetic Peptide, PEP-1026, from Invitrogen.
CAZIP, also known as TIP150, contains a leucine-zipper domain at the C-terminus and has at least three isoforms. CAZIP is expressed in heart during early cardiac development and in parts of the nervous system in later embryonic development. It is thought to bind microtubules and facilitate the end-binding protein 1 (EB1)-dependent loading of MCAK onto microtubules plus ends and orchestrates the dynamics at the plus end of microtubules. CAZIP may regulate the dynamics of microtubules at their growing distal tip.
Specifications
Specifications
| Gene ID (Entrez) | 23281 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q5JR59 |
| Common Name | Human CAZIP |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction