missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin V Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB1080-SP
This item is not returnable.
View return policy
Description
Cathepsin V Monoclonal specifically detects Cathepsin V in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Cathepsin V | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
| O60911 | |
| CTSL2 | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin V Accession # O60911 | |
| 25 μg | |
| Primary | |
| Detects human Cathepsin V in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Cathepsin H and recombinant human (rh) Cathepsin K is observed and no cross-reactivity with rhCathepsin B, C, L, O, S, Z, rmCathepsin B, C, F, or Z is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 182016 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cathepsin L2, Cathepsin U, Cathepsin V, CATL2, CTSL2, CTSUcathepsin L2, preproprotein, CTSVMGC125957, EC 3.4.22, EC 3.4.22.43 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1515 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction