missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin F Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2075
This item is not returnable.
View return policy
Description
Cathepsin F Polyclonal specifically detects Cathepsin F in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Cathepsin F | |
| Polyclonal | |
| Unconjugated | |
| Q9UBX1 | |
| CTSF | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin F Ala20-Asp484 Accession # Q9UBX1 | |
| 100 μg | |
| Primary | |
| Detects human Cathepsin F in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Cathepsin F is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| cathepsin F, CATSFEC 3.4.22.41, CTSF, EC 3.4.22 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8722 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction