missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin E Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
£168.00 - £432.00
Specifica
| Antigen | Cathepsin E |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
18735387
![]() |
R&D Systems
AF1294-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18069321
![]() |
R&D Systems
AF1294 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Cathepsin E Polyclonal specifically detects Cathepsin E in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifica
| Cathepsin E | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
| CTSE | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| NP_001901 | |
| 1510 | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin E Ile54-Pro396 Accession # NP_001901 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human pro and active Cathepsin E in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse Cathepsin E is observed and less than 2% cross-reactivity with recombinant human (rh) Cathepsin D and rhBACE is observed. |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto