missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cathepsin C ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human Cathepsin C quantitates human Cathepsin C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cathepsin C.
Cathepsin C, known also as dipeptidyl aminopeptidase I (DPPI), is a tetrameric lysosomal cysteine peptidase belonging to the papain family. Cathepsin C is involved in intracellular protein degradation and the processing of protein precursors, where it participates in cell growth, neuraminidase activation, and platelet factor XIII activation. Cathepsin C is largely related to other lysosomal cysteine proteinases, including cathepsin B, H and L. Enzymatically, Cathepsin C is capable of sequentially removing dipeptides from the amino terminus, and it requires halide ions, namely chloride ions, and thiols for complete enzymatic activity. Protein levels of Cathepsin C are detected in a variety of tissues, and it is most highly expressed in spleen, kidney, cytotoxic lymphocytes and myeloid cells, where it localizes to the secretory granule compartment. Cathepsin C is initially synthesized as a proenzyme that is rapidly processed to generate two distinct chains that function together as the mature form of the enzyme.
Specifications
Specifications
| Accession Number | AI047818; CATC; Cathepsin C; cathepsin J; CPPI; CTSC; dipeptidyl peptidase 1; Dipeptidyl peptidase 1 exclusion domain chain; Dipeptidyl peptidase 1 heavy chain; Dipeptidyl peptidase 1 light chain; Dipeptidyl peptidase I; Dipeptidyl peptidase I exclusion domain chain; Dipeptidyl peptidase I heavy chain; Dipeptidyl peptidase I light chain; Dipeptidyl transferase; dipeptidyl-peptidase; dipeptidylpeptidase 1; dipeptidyl-peptidase 1; dipeptidyl-peptidase I; DPP1; DPPI; DPP-I; HMS; JP; JPD; PALS; PDON1; PLS |
| Assay Range | 2 to 500 ng/mL |
| Assay Sensitivity | 2 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 1075 |
| Gene Symbol | CTSC |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction