missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cathelicidin ELISA Kit

Product Code. 30245074
Click to view available options
:
96 Tests
This item is not returnable. View return policy

Product Code. 30245074

Brand: Invitrogen™ EEL201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Cathelicidin is an epithelial-derived antimicrobial protein found in specific granules of polymorphonuclear leukocytes of bone marrow, testis and neutrophils. Cathelicidins are one family of antimicrobial peptides characterized by conserved pro-peptide sequences that have been identified in epithelial tissues and some myeloid cells of humans and animals. LL-37/hCAP-18 is the only Cathelicidin found in humans and is expressed in inflammatory and epithelial cells. The presence of these molecules is essential for defense against invasive bacterial infection in skin. Besides their direct antimicrobial function, Cathelicidins have multiple roles in mediating innate and adaptive immunity, such as endotoxin neutralizing, angiogenesis, wound healing and promoting neutrophil chemotaxis and mast cell recruitment. Cathelicidin antimicrobial peptides qualify as prototypes of innovative drugs that may be used to treat infection and/or modulate the immune response.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49913
Assay Range 1.56 to 100 ng/mL
Assay Sensitivity 0.94 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format ELISA Microplate (Dismountable)
Product Type ELISA
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 820
Gene Symbol CAMP
Interassay CV <10
Intraassay CV <10
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Cathelicidin
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.