missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caspr1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7548-SP
This item is not returnable.
View return policy
Description
Caspr1 Monoclonal specifically detects Caspr1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Caspr1 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P78357 | |
| CNTNAP1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Caspr1 Trp20-Gly1280 Accession # P78357 | |
| 25 μg | |
| Primary | |
| Detects human Caspr1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Caspr2 is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 753232 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Caspr, Caspr1, CASPRcaspr1, CNTNAP, CNTNAP1, contactin associated protein 1, neurexin 4, Neurexin IV, Neurexin-4, NRXN4contactin-associated protein 1, P190 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8506 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction