Learn More
Invitrogen™ Caspase 8/FLICE Human ELISA Kit
For the quantitative detection of human Caspase-8.
Brand: Invitrogen™ BMS2024TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human Caspase-8
anti-human Caspase-8 polyclonal (rabbit) Detection Antibody
Anti-rabbit-IgG-HRP
Human Caspase-8 Standard lyophilized, 20.00ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Lysis Buffer 10x
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Description
The Human Caspase 8/FLICE ELISA quantitates Hu Caspase 8 in human serum, cell lysate or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Caspase 8. Principle of the method The Human Caspase 8 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Caspase 8 (CASP8) is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases play a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes composed of a pro-domain, a large protease subunit, and a small protease subunit. Activation of caspases requires proteolytic processing at conserved internal aspartic residues to generate a heterodimeric enzyme consisting of the large and small subunits. Caspase 8 is involved in the programmed cell death induced by Fas and various apoptotic stimuli. The N-terminal FADD-like death effector domain of Caspase 8 suggests that it may interact with Fas-interacting protein FADD. Caspase 8 was detected in the insoluble fraction of the affected brain region from Huntington disease patients but not in those from normal controls, which implicated the role in neurodegenerative diseases. Caspase 8 binds to the death effector domain (DED) of FADD through an analogous DED domain present in tandem in the pro-form of the Caspase 8 protein. Activated Caspase 8 then activates other downstream caspases including Caspase 9, thereby committing the cell to undergo apoptosis. In addition, Caspase 8 also reacts with Jurkat cells and Tonsil. Overexpression of Caspase 8 induces apoptosis, which can be blocked by inhibitors specific for the ICE family. Many alternatively spliced transcript variants encoding different isoforms have been described for Caspase 8, however, not all variants have had their full-length sequences determined.Specifications
Q14790 | |
0.10 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
ALPS2B,CAP4,Casp-8,FLICE,MACH,MCH5 | |
6.7% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 15 min. |
0.16-10.0 ng/mL | |
HRP | |
Cell Lysates, Serum, Supernatant | |
ELISA | |
841 | |
8.5% | |
Pre-coated 96 well plate, Standard, Standard, Sample Diluent, Assay Buffer concentrate, Detection Antibody, anti-rabbit HRP conjugate, Wash Buffer, Wash Buffer, Stop Solution, Stop Solution | |
10 x 96 Tests | |
Cell Lysate,50 μL; Serum, 50 μL; Supernatant, 50 μL | |
MACH, FADD-homologous ICE/CED-3-like protease | |
1 hr. 20 min. |
For Research Use Only