missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Caspase 5 (aa 7-92) Control Fragment Recombinant Protein

Product Code. 30181967
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181967

Brand: Invitrogen™ RP98453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (23%), Rat (23%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111126 (PA5-111126. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12. Activation of these caspases results in the cleavage and activation of proinflammatory cytokines such as IL-1-b and IL-18. Caspase-5 can interact with caspase-1; both are constituents of the NALP1 inflammasome, a complex that can trigger the cleavage of pro-IL-1-b. Expression of caspase-5 can be regulated by lipopolysaccharide (LPS) and IFN-gamma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51878
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 838
Name Human Caspase 5 (aa 7-92) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CASP5; CASP-5; caspase 5; caspase 5, apoptosis-related cysteine peptidase; caspase 5, apoptosis-related cysteine protease; caspase-5; Caspase-5 subunit p10; Caspase-5 subunit p20; ICE(rel)III; ICE(rel)-III; ICEREL-III; ICH3; ICH-3; Protease ICH-3; protease TY; TY protease
Common Name Caspase 5
Gene Symbol CASP5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VEGVFIFLIEDSGKKKRRKNFEAMFKGILQSGLDNFVINHMLKNNVAGQTSIQTLVPNTDQKSTSVKKDNHKKKTVKMLEYLGKDV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.