missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caspase-10 (FLICE 2) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF834-SP
This item is not returnable.
View return policy
Description
Caspase-10/FLICE2 Polyclonal specifically detects Caspase-10/FLICE2 in Human samples. It is validated for Western Blot.
Specifications
| Caspase-10/FLICE2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CASP10 | |
| KLH-coupled human Caspase-10 (FLICE 2) synthetic peptide KRTVWGAKQISATSLPTAC | |
| 25 μg | |
| Primary | |
| Detects human and mouse Caspase-10 (FLICE 2) precursor in Western blots. Sensitivity for detection of the small subunit is not known. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Apoptotic protease Mch-4, CASP10, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, Caspase10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, Mch4, MCH4ALPS2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 843 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur