missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Caspase-1/ICE Quantikine ELISA Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
Description
The Human Caspase-1/ICE Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Caspase-1 in cell culture supernates (100 ul).
Specifications
Specifications
| Assay Range | 6.25 - 400pg/mL |
| Assay Sensitivity | 1.24pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant |
| Specificity | Natural and Recombinant Human Caspase-1 |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 834 |
| Sample Volume | Cell Culture Supernates (100uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction