missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caspase-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB62152-100
This item is not returnable.
View return policy
Description
Caspase-1 Monoclonal antibody specifically detects Caspase-1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
Specifications
| Caspase-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 834 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunocytochemistry | |
| 1021323 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| CASP1, CASP-1, CASP1 nirs variant 1, caspase 1, apoptosis-related cysteine peptidase, caspase-1, Caspase1, Caspase-1 subunit p10, Caspase-1 subunit p20, EC 3.4.22.36, EC:3.4.22.36, ICE, IL-1 beta-converting enzyme, IL1BC, IL-1BC, IL1BCE, IL1B-convertase, interleukin 1, beta, convertase, interleukin 1-B converting enzyme, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45 | |
| Human Caspase-1 peptide | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction