missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carnosine Dipeptidase 1/CNDP1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2489
This item is not returnable.
View return policy
Description
Carnosine Dipeptidase 1/CNDP1 Polyclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.
Specifications
| Carnosine Dipeptidase 1/CNDP1 | |
| Polyclonal | |
| Unconjugated | |
| Q96KN2 | |
| CNDP1 | |
| Mouse myeloma cell line NS0-derived recombinant human Carnosine Dipeptidase 1/CNDP1 Pro28-His507 Accession # Q96KN2 | |
| 100 μg | |
| Primary | |
| Detects human Carnosine Dipeptidase 1/CNDP1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CNDP1, CPGL2, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84735 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction