missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CARF Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4195-SP
This item is not returnable.
View return policy
Description
CARF/CDKN2AIP Polyclonal specifically detects CARF/CDKN2AIP in Human samples. It is validated for Western Blot.
Specifications
| CARF/CDKN2AIP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CARFCDKN2A-interacting protein, CDKN2A interacting protein, CDKN2AIP, collaborates/cooperates with ARF (alternate reading frame) protein, Collaborator of ARF, FLJ20036 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55602 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NXV6 | |
| CDKN2AIP | |
| E. coli-derived recombinant human CARF Ser379-Leu579 Accession # Q9NXV6 | |
| 25 μg | |
| Primary | |
| Detects human CARF in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction