missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cardiotrophin-1/CT-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £1455.00
Specifications
| Antigen | Cardiotrophin-1/CT-1 |
|---|---|
| Clone | 89215 |
| Dilution | Western Blot 1 ug/mL, Neutralization 1-3 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18769527
![]() |
R&D Systems
MAB2601-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755699
![]() |
R&D Systems
MAB2601-100 |
100 μg |
£361.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18734998
![]() |
R&D Systems
MAB2601-500 |
500 μg |
£1455.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for Western Blot, Neutralization.Specifications
| Cardiotrophin-1/CT-1 | |
| Western Blot 1 ug/mL, Neutralization 1-3 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q16619 | |
| 1489 | |
| E. coli-derived recombinant human Cardiotrophin-1/CT-1 Ser2-Ala201 Accession # Q16619 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Cardiotrophin-1/CT-1 in direct ELISAs and Western blots. |
| 89215 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1 | |
| CTF1 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title