missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cardiac Myoglobin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems G-125-C
This item is not returnable.
View return policy
Description
Myoglobin Polyclonal specifically detects Myoglobin in Human samples. It is validated for .
Specifications
| Myoglobin | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and Sodium Azide with Trehalose. with No Preservative | |
| MB | |
| Human cardiac Myoglobin | |
| 0.1 mg | |
| Primary | |
| This antibody reacts with cardiac and skeletal forms of myoglobin. | |
| Human | |
| IgG |
| Polyclonal | |
| Hidden | |
| MB, MGC13548, myoglobin, PVALB | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 4151 | |
| Reconstitute with sterile PBS. If 0.5 mL of PBS is used, the antibody concentration will be 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction