missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carboxypeptidase A1/CPA1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2856
This item is not returnable.
View return policy
Description
Carboxypeptidase A1/CPA1 Polyclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
Specifications
| Carboxypeptidase A1/CPA1 | |
| Polyclonal | |
| Unconjugated | |
| P15085 | |
| CPA1 | |
| Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase A1/CPA1 Lys17-Tyr419 Accession # P15085 | |
| 100 μg | |
| Primary | |
| Detects human Carboxypeptidase A1/CPA1 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse CPA1 is observed and less than 10% cross-reactivity with recombinant human CPB1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.7 ug/mL | |
| carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, CPA1, EC 3.4.17, EC 3.4.17.1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1357 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction