missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carboxylesterase 2/CES2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5657
This item is not returnable.
View return policy
Description
Carboxylesterase 2/CES2 Monoclonal specifically detects Carboxylesterase 2/CES2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.
Specifications
| Carboxylesterase 2/CES2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O00748 | |
| CES2 | |
| Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase 2/CES2 Gln27-Leu559 (predicted) Accession # O00748 | |
| 100 μg | |
| Primary | |
| Detects human Carboxylesterase 2/CES2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Carboxylesterase 2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 720711 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| carboxylesterase 2 (intestine, liver), carboxylesterase 2EC 3.1.1.1, CE-2, CE-2PCE-2, CES2, CES2A1, EC 3.1.1, EC 3.1.1.84, hCE-2, iCE, intestinal carboxylesterase; liver carboxylesterase-2, PCE-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8824 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction