missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase XII/CA12 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2190
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XII/CA12 Monoclonal specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Western Blot.
Specifications
| Carbonic Anhydrase XII/CA12 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O43570 | |
| CA12 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase XII/CA12 Ala25-Gln291 Accession # O43570 | |
| 100 μg | |
| Primary | |
| Detects human Carbonic Anhydrase XII/CA12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CA1, -2, -3, -4, -7, -8, -10, -11, -13 or -14 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 315602 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CA12, Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 771 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction