missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase IV/CA4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB21861
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IV/CA4 Monoclonal specifically detects Carbonic Anhydrase IV/CA4 in Human samples. It is validated for Western Blot.
Specifications
| Carbonic Anhydrase IV/CA4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P22748 | |
| CA4 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IV/CA4 Ala19-Lys283 Accession # P22748 | |
| 100 μg | |
| Primary | |
| Detects human Carbonic Anhydrase IV/CA4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CA1, -2, -3, -5a, -5b, -7, 8, -9, or -10 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 310415 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CA4, CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant), RP17 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 762 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction