missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Carbonic Anhydrase III Recombinant Protein

Recombinant Protein

Brand:  Invitrogen™ RP75525

Code : LD

Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg

Product Code. 11594211

  • £602.00 / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CAIII (human) Recombinant Protein, RP-75525, from Invitrogen.

This product contains purified recombinant human CAIII protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
Specifications

Specifications

P07451
PBS with 50% glycerol and no preservative
Human Carbonic Anhydrase III
E. coli
-20°C, Avoid Freeze/Thaw Cycles
Carbonic Anhydrase III
CA3
Unconjugated
E. coli
Liquid
purified
Control, Western Blot
761
100μg
N/A
RUO
Human Carbonic Anhydrase III
Human
Hu
Recombinant
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers