missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase III/CA3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2185-SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase III/CA3 Monoclonal specifically detects Carbonic Anhydrase III/CA3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Carbonic Anhydrase III/CA3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
| P07451 | |
| CA3 | |
| E. coli-derived recombinant human Carbonic Anhydrase III/CA3 Ala2-Lys260 Accession # P07451 | |
| 25 μg | |
| Primary | |
| Detects human Carbonic Anhydrase III/CA3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CA1, 2, 4, 5a, 5b, 6, 7, 8, or 9 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 495018 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CA3, CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 761 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction