missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase I/CA1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2180-SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase I/CA1 Monoclonal specifically detects Carbonic Anhydrase I/CA1 in Human samples. It is validated for Western Blot.
Specifications
| Carbonic Anhydrase I/CA1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P00915 | |
| CA1 | |
| E. coli-derived recombinant human Carbonic Anhydrase I/CA1 Ala2-Phe261 Accession # P00915 | |
| 25 μg | |
| Primary | |
| Detects human Carbonic Anhydrase I/CA1 in direct ELISAs and Western blots. In Western blots, 30% cross-reactivity with recombinant human (rh) CA2 is observed and no cross-reactivity with rhCA3, -4, -7, -8, -10, -11, -12, -13, or -14 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 363121 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CA1, CA-I, Car1, Carbonate dehydratase I, carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase ICAB, carbonic dehydratase, EC 4.2.1.1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 759 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction