missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbohydrate Sulfotransferase 5/CHST5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5446-SP
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 5/CHST5 Polyclonal specifically detects Carbohydrate Sulfotransferase 5/CHST5 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Carbohydrate Sulfotransferase 5/CHST5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5, CHST5, GlcNAc6ST3, gn6st-3, GST4-alpha, hIGn6ST, I-GlcNAc6ST | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23563 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9GZS9 | |
| CHST5 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Carbohydrate Sulfotransferase 5/CHST5 Ser28-Arg390 Accession # Q9GZS9 | |
| 25 μg | |
| Primary | |
| Detects human Carbohydrate Sulfotransferase 5/CHST5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) CHST1, rhCHST2, rmCHST5, and rmCHST7 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction