missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Calreticulin ELISA Kit
Description
Human Calreticulin quantitates human Calreticulin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Calreticulin.
Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
Specifications
Specifications
| Accession Number | Autoantigen RO; CABP3; CALBP; Calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99 n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99 n; I79 |
| Assay Range | 1.3 to 200 ng/mL |
| Assay Sensitivity | 1.3 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 811 |
| Gene Symbol | CALR |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?