missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Calponin 1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7900
This item is not returnable.
View return policy
Description
Calponin 1 Monoclonal specifically detects Calponin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Calponin 1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P51911 | |
| CNN1 | |
| E. coli-derived recombinant human Calponin 1 Ser2-Ala297 Accession # P51911 | |
| 100 μg | |
| Primary | |
| Detects human Calponin 1 in ELISAs. Detects human, mouse and rat Calponin 1 in Western Blots. In direct ELISAs, no cross-reactivity with recombinant human Calponin 3 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 836701 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, Calponin H1, smooth muscle, calponins, basic, CNN1, Sm-Calp, SMCC, SMCCcalponin-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1264 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction