missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Calpain 11 (aa 547-629) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (59%), Rat (59%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83168 (PA5-83168. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
Specifications
Specifications
| Accession Number | Q9UMQ6 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 11131 |
| Name | Human Calpain 11 (aa 547-629) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | calcium-activated neutral proteinase 11; calcium-activated neutral proteinase 11 {ECO:0000250; calcium-dependent thiol protease; calpain 11; calpain 11 {ECO:0000312; calpain11; Calpain-11; calpain-11; LOW QUALITY PROTEIN: calpain-11; CANP 11; CANP 11 {ECO:0000250; Capn11; capn11 {ECO:0000312; EMBL:AAH97256.1}; HIP2; LIG; RGD:1302946}; UniProtKB:Q9UMQ6} |
| Common Name | Calpain 11 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction