Learn More
Human Caldesmon/CALD1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB7569-SP
Product Code. 18799697
  • £146.00 / 25µg
Description
Caldesmon/CALD1 Monoclonal specifically detects Caldesmon/CALD1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caldesmon/CALD1
Monoclonal
LYOPH
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Q05682
CALD1
E. coli-derived recombinant human Caldesmon/CALD1 Lys696-Val793 Accession # Q05682
25 μg
Primary
Detects human Caldesmon/CALD1 in direct ELISAs and Western blots.
Human
Purified
Western Blot, Immunocytochemistry
767001
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CAD, CALD1, caldesmon, caldesmon 1, CDM, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
800
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b