missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Calcitonin R Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4614G-100UG
This item is not returnable.
View return policy
Description
Calcitonin R Monoclonal specifically detects Calcitonin R in Human samples. It is validated for Flow Cytometry.
Specifications
| Calcitonin R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_001733 | |
| CALCR | |
| HEK293 human embryonic kidney cell line transfected with human Calcitonin R Ala25-Ala474 Accession # NP_001733 | |
| RUO | |
| 799 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 462802 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| calcitonin receptor, CalcitoninR, CALCR, CRT, CTR, CT-R, CTR1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Calcitonin R. Stains human Calcitonin R transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction