missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Calbindin D Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3320-SP
This item is not returnable.
View return policy
Description
Calbindin D/S100G Monoclonal specifically detects Calbindin D/S100G in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Calbindin D/S100G | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P05937 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 795 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 401025 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| S100G | |
| E. coli-derived recombinant human Calbindin D Met1-Asn261 Accession # P05937 | |
| 25 μg | |
| Primary | |
| Detects human Calbindin D in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human S100P. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction