missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cadherin-6/KCAD Alexa Fluor 350 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2715U-100UG
This item is not returnable.
View return policy
Description
Cadherin-6/KCAD Monoclonal specifically detects Cadherin-6/KCAD in Human samples. It is validated for Flow Cytometry.
Specifications
| Cadherin-6/KCAD | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P55285 | |
| CDH6 | |
| Mouse myeloma cell line NS0-derived recombinant human Cadherin-6/KCAD Ser54-Ala615 Accession # P55285 | |
| RUO | |
| 1004 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 427909 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| cadherin 6, type 2, K-cadherin (fetal kidney), cadherin, fetal kidney, cadherin-6, Cadherin6, CDH6, KCAD, K-Cadherin, K-cadherin (fetal kidney) | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Cadherin-6/KCAD in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cadherin-8, -11, -12, -13, -17, rhECAD, rhMCAD, rhNCAD, rhRCAD, rhPCAD, or rhVECAD is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction