missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cadherin-13 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3264
This item is not returnable.
View return policy
Description
Cadherin-13 Polyclonal specifically detects Cadherin-13 in Human samples. It is validated for Western Blot.
Specifications
| Cadherin-13 | |
| Polyclonal | |
| Biotin | |
| P55290 | |
| CDH13 | |
| Mouse myeloma cell line NS0-derived recombinant human Cadherin-13 Glu23-Ala692 Accession # P55290 | |
| 50 μg | |
| Primary | |
| Detects human Cadherin-13 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-11, rhCadherin-17, rhE-Cadherin, rhP-Cadherin, rhVE-Cadherin, and rhR-Cadherin is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| cadherin 13, H-cadherin (heart), cadherin-13, Cadherin13, CDH13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, T-Cadherin, Truncated cadherin, truncated-cadherin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1012 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction