missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CACNA1F (aa 1813-1889) Control Fragment Recombinant Protein

Product Code. 30205912
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205912

Brand: Invitrogen™ RP108540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84778 (PA5-84778. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Voltage-sensitive calcium channels (VSCC) mediate the entry of calcium ions into excitable cells and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. The isoform alpha-1F gives rise to L-type calcium currents. Long-lasting (L-type) calcium channels belong to the 'high-voltage activated' (HVA) group. They are blocked by dihydropyridines (DHP), phenylalkylamines, benzothiazepines, and by omega-agatoxin-IIIA (omega-Aga-IIIA). They are however insensitive to omega-conotoxin-GVIA (omega-CTx-GVIA) and omega-agatoxin-IVA (omega-Aga-IVA).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60840
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 778
Name Human CACNA1F (aa 1813-1889) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A930034B14; AIED; Cacna1f; CACNAF1; calcium channel, voltage-dependent, alpha 1 F subunit; calcium channel, voltage-dependent, L type, alpha 1 F subunit; calcium voltage-gated channel subunit alpha1 F; Cav1.4; Cav1.4 alpha1; COD3; COD4; CORDX; CORD x 3; CSNB2; CSNB2A; CSNB x 2; JM8; JMC8; L-type dihydropyridine-sensitive calcium channel alpha-1 f subunit; nerg1; nob2; OA2; Sfc17; Voltage-dependent L-type calcium channel subunit alpha-1 F; voltage-gated calcium channel subunit alpha Cav1.4
Common Name CACNA1F
Gene Symbol CACNA1F
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SCEDLPIPGTYHRGRNSGPNRAQGSWATPPQRGRLLYAPLLLVEEGAAGEGYLGRSSGPLRTFTCLHVPGTHSDPSH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.