missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CABYR (aa 181-273) Control Fragment Recombinant Protein

Product Code. 30182394
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182394

Brand: Invitrogen™ RP98250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (55%), Rat (55%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59256 (PA5-59256. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Spermatozoa gain fertilization capacitation and hyperactivation after residing in the uterus or oviduct. Calcium, cAMP and protein tyrosine phosphorylation are necessary for the molecular mechanisms that allow for this transformation. CABYR (Calcium-binding tyrosine phosphorylation-regulated protein), also known as Fibrousheathin-2 or Cancer/testis antigen 88, is a 493 amino acid protein that is expressed in sperm flagella and exhibits increased tyrosine phosphorylation during capacitation. There are six named isoforms of CABYR that are produced as a result of alternative slicing events. Specifically, isoform 1 is expressed in the testis, while isoform 3 and isoform 5 are expressed in pancreas, brain and various brain tumors. Isoforms 1, 2 and 6 most likely bind calcium, whereas isoforms 3, 4 and 5 probably do not bind calcium.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75952
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26256
Name Human CABYR (aa 181-273) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700016C01Rik; 4933421A18Rik; Cabyr; CABYRa; CABYRc; CABYRc/d; CABYRe; calcium binding tyrosine phosphorylation regulated; calcium binding tyrosine-(Y)-phosphorylation regulated; calcium binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); calcium-binding protein 86; Calcium-binding tyrosine phosphorylation-regulated protein; calcium-binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); calcium-binding tyrosine-phosphorylation regulated protein; cancer/testis antigen 88; CBP86; CT88; fibrousheathin 2; fibrousheathin II; Fibrousheathin-2; FSP2; FSP-2; testis tissue sperm-binding protein Li 84 P; testis-specific calcium-binding protein CBP86
Common Name CABYR
Gene Symbol CABYR
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GKVSSIHSDQSDVLMVDVATSMPVVIKEVPSSEAAEDVMVAAPLVCSGKVLEVQVVNQTSVHVDLGSQPKENEAEPSTASSVPLQDEQEPPAY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.