missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C9orf156 (aa 328-441) Control Fragment Recombinant Protein

Product Code. 30208724
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208724

Brand: Invitrogen™ RP92976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54368 (PA5-54368. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

S-adenosyl-L-methionine-dependent methyltransferase responsible for the addition of the methyl group in the formation of N6-methyl-N6-threonylcarbamoyladenosine at position 37 (m(6)t(6)A37) of the tRNA anticodon loop of tRNA(Ser)(GCU) (PubMed:25063302). The methyl group of m(6)t(6)A37 may improve the efficiency of the tRNA decoding ability. May bind to tRNA. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BU70
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51531
Name Human C9orf156 (aa 328-441) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5830415F09Rik; AV014846; C9orf156; HSPC219; likely ortholog of H. sapiens chromosome 9 open reading frame 156 (C9orf156); NAP1; Nef (lentivirus myristoylated factor) associated protein 1; Nef associated protein 1; nef-associated protein 1; RGD1305420; RIKEN cDNA 5830415F09 gene; similar to Nef associated protein 1; thioesterase NAP1; TRMO; tRNA (adenine(37)-N6)-methyltransferase; tRNA methyltransferase O
Common Name C9orf156
Gene Symbol TRMO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence WVTEAPVATLEVRFTPHAEMDLGQLSSQDVGQASFKYFQSAEEAKRAIEAVLSADPRSVYRRKLCQDRLFYFTVDIAHVTCWFGDGFAEVLRIKPASEPVHMTGPVGSLVSLGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.