missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human C5a Recombinant Protein, PeproTech®

Product Code. 17822273
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17822273

Brand: Gibco™ 30070100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

300-70-1MG will be provided as 2 x 500 μg (300-70-500UG). Recombinant Human C5a is an 8.3 kDa glycoprotein containing the 74 amino acid residues of the C5a anaphylatoxin chain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is the fifth component of complement, which plays an important role in inflammatory and cell killing processes. This protein is comprised of alpha and beta polypeptide chains that are linked by a disulfide bridge. An activation peptide, C5a, which is an anaphylatoxin that possesses potent spasmogenic and chemotactic activity, is derived from the alpha polypeptide via cleavage with a convertase. The C5b macromolecular cleavage product can form a complex with the C6 complement component, and this complex is the basis for formation of the membrane attack complex, which includes additional complement components. Mutations in this gene cause complement component 5 deficiency, a disease where patients show a propensity for severe recurrent infections. Defects in this gene have also been linked to a susceptibility to liver fibrosis and to rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01031
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 727
Molecular Weight (g/mol) 8.3 kDa
Name Human C5a
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905
Common Name Complement C5a
Gene Symbol C5
Biological Activity Measured by its ability to support adhesion of Human Umbilical Vein Endothelial Cells (HUVEC) to the Recombinant Human C5a-coated surface.
Conjugate Unconjugated
Recombinant Recombinant
Sequence TLQKKIEEIA AKYKHSVVKK CCYDGACVNN DETCEQRAAR ISLGPRCIKA FTECCVVASQ LRANISHKDM QLGR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.