Learn More
Invitrogen™ Complement C5a Human ELISA Kit
Sandwich ELISA Kit
£451.00 - £3070.00
Specifications
Target Species | Human |
---|---|
For Use With (Application) | ELISA |
Synonym | complement component 5, C5a, C5D, C5b, ECLZB, CPAMD4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15314988
|
Invitrogen™
BMS2088 |
96 Tests |
£451.00
1 set |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15501617
|
Invitrogen™
BMS2088TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Description
The Human Complement C5a (Hu C5a) ELISA quantitates Hu C5a in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu C5a. Principle of the method The Human C5a solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
Human | |
ELISA | |
complement component 5, C5a, C5D, C5b, ECLZB, CPAMD4 |