Learn More
Invitrogen™ C1q Human ELISA Kit
For the quantitative detection of human C1q.
Brand:  Invitrogen™ BMS2099FIVE
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15511657
  • £1548.00 / Pack of 5
In Stock
to see stock.
Add to basket
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human C1q
Description
Description
The Human C1q ELISA quantitates Hu C1q in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu C1q. Principle of the method The Human C1q solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
0.08ng/mL
Human
5 x 96 Tests
C1q
Cell Culture Supernatant, Plasma, Serum
ELISA
complement component 1, q subcomponent, A chain el, complement component 1, q subcomponent, alpha poly