missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C1orf96 (aa 192-270) Control Fragment Recombinant Protein

Product Code. 30198277
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198277

Brand: Invitrogen™ RP94330

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55838 (PA5-55838. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plays a role in microtubule (MT) stabilization and this stabilization involves the maintenance of NUMA1 at the spindle poles. Colocalizes with polyglutamylated MTs to promote MT stabilization and regulate bipolar spindle formation in mitosis. Binding of CCSAP to centrosomes and the spindle around centrosomes during mitosis inhibits MT depolymerization, thereby stabilizing the mitotic spindle (PubMed:26562023). May play a role in embryonic development. May be required for proper cilia beating. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6IQ19
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 126731
Name Human C1orf96 (aa 192-270) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700054N08Rik; AI853424; C1orf96; C4H1orf96; Ccsap; centriole and spindle-associated protein; centriole, cilia and spindle associated protein; centriole, cilia and spindle-associated protein; CSAP; RGD1563235
Common Name C1orf96
Gene Symbol CCSAP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KQTDTGSQKTHNVCASAPVHEIHESALRAKNRRQVEKRKLVAQRQRAHSVDVEKNRKMKASSSENPWMTEYMRCYSARA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.