missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C1orf86 (aa 108-180) Control Fragment Recombinant Protein

Product Code. 30209738
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209738

Brand: Invitrogen™ RP94374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (58%), Rat (58%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58555 (PA5-58555. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

C1orf86 is a component of the Fanconi anemia (FA) complex required to recruit the FA complex to DNA interstrand cross-links (ICLs) and promote ICLs repair. Following DNA damage recognizes and binds 'Lys-63'-linked ubiquitin generated by RNF8 at ICLs and recruits other components of the FA complex. It promotes translesion synthesis via interaction with REV1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6NZ36
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 199990
Name Human C1orf86 (aa 108-180) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610002J02Rik; C1orf86; Faap20; FANCA-associated protein of 20 kDa; Fanconi anemia core complex associated protein 20; Fanconi anemia core complex-associated protein 20; Fanconi anemia-associated protein of 20 kDa; FP7162; likely orthologue of H. sapiens chromosome 1 open reading frame 86 (C1orf86); LOC362678; RGD1308923; RIKEN cDNA 2610002J02 gene
Common Name C1orf86
Gene Symbol FAAP20
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GGHLESPARSLPQRPAPDPCRAPRVEQQPSVEGAAALRSCPMCQKEFAPRLTQLDVDSHLAQCLAESTEDVTW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.