missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human C1GALT1C1 (aa 34-152) Control Fragment Recombinant Protein

Product Code. 30207932
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207932

Brand: Invitrogen™ RP92239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53349 (PA5-53349. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a type II transmembrane protein that is similar to the core 1 beta1,3-galactosyltransferase 1, which catalyzes the synthesis of the core-1 structure, also known as Thomsen-Friedenreich antigen, on O-linked glycans. This gene product lacks the galactosyltransferase activity itself, but instead acts as a molecular chaperone required for the folding, stability and full activity of the core 1 beta1,3-galactosyltransferase 1. Mutations in this gene have been associated with Tn syndrome. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96EU7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29071
Name Human C1GALT1C1 (aa 34-152) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500002I11Rik; AB030184; beta 1,3-galactosyltransferase 2; beta1,3-galactosyltransferase 2; C1GALT1 specific chaperone 1; C1galt1c1; C1GALT1-specific chaperone 1; C1GALT2; C1Gal-T2; C38H2-L1; C38H2-like protein 1; C81205; core 1 beta1,3-galactosyltransferase 2; core 1 beta3-galactosyltransferase-specific molecular chaperone; core 1 beta3-Gal-T2; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 2; COSMC; HSPC067; mC1Gal-T2; MST143; MSTP143; RGD1311230; TNPS; UNQ273/PRO310
Common Name C1GALT1C1
Gene Symbol C1GALT1C1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NRMHHHEHHHLQAPNKEDILKISEDERMELSKSFRVYCIILVKPKDVSLWAAVKETWTKHCDKAEFFSSENVKVFESINMDTNDMWLMMRKAYKYAFDKYRDQYNWFFLARPTTFAIIE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado